missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzamide, 98+%, Thermo Scientific Chemicals

Product Code. 11470213
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470213 50 g 50g
11460213 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470213 Supplier Thermo Scientific Alfa Aesar Supplier No. A11389.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-48-4
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD00014799
InChI Key MJTGQALMWUUPQM-UHFFFAOYSA-N
Synonym m-chlorobenzamide, benzamide, 3-chloro, 3-chloro-benzamide, benzamide, m-chloro-8ci, benzamide, m-chloro, 5-chlorobenzamide, pubchem3618, acmc-1b3cu, wce
PubChem CID 69254
ChEBI CHEBI:10587
IUPAC Name 3-chlorobenzamide
SMILES NC(=O)C1=CC=CC(Cl)=C1

Specifications

Melting Point 133°C to 136°C
Quantity 50 g
Beilstein 1859940
Formula Weight 155.58
Percent Purity ≥98%
Chemical Name or Material 3-Chlorobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.