Learn More
3-chlorobenzaldehyde, 99%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 587-04-2 |
---|---|
Molecular Formula | C7H5ClO |
Molecular Weight (g/mol) | 140.57 |
MDL Number | MFCD00003350 |
InChI Key | SRWILAKSARHZPR-UHFFFAOYSA-N |
Synonym | m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde |
PubChem CID | 11477 |
IUPAC Name | 3-chlorobenzaldehyde |
SMILES | C1=CC(=CC(=C1)Cl)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10132470
|
Thermo Scientific Acros
108610250 |
25 mL | Glass bottle |
N/A
|
N/A | ||||
10786511
|
Thermo Scientific Acros
108611000 |
100 mL | Glass bottle |
N/A
|
N/A | ||||
10686354
|
Thermo Scientific Acros
108615000 |
500 mL | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
587-04-2 | |
140.57 | |
SRWILAKSARHZPR-UHFFFAOYSA-N | |
11477 | |
C1=CC(=CC(=C1)Cl)C=O |
C7H5ClO | |
MFCD00003350 | |
m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde | |
3-chlorobenzaldehyde |
Specifications
587-04-2 | |
100.0 | |
Colorless to Yellow | |
213°C to 215°C | |
Authentic | |
C7H5ClO | |
1.5630 to 1.565 | |
07, 234 | |
m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde | |
SRWILAKSARHZPR-UHFFFAOYSA-N | |
3-chlorobenzaldehyde | |
11477 | |
99% | |
3-chlorobenzaldehyde |
98.5 | |
17°C to 18°C | |
1.2410g/mL | |
88°C | |
98.5% min. (GC) | |
ClC6H4CHO | |
MFCD00003350 | |
1.241 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and acetone | |
C1=CC(=CC(=C1)Cl)C=O | |
140.57 | |
140.57 | |
Liquid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-596-9
- 3-Chlorobenzaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only