missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloroaniline, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
3-Chloroaniline was the only aniline compound to increase plasmaALT/GPT activity at 48 h. It is employed in the bioaugmentation of activated sludge.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chloroaniline was the only aniline compound to increase plasmaALT/GPT activity at 48 h. It is employed in the bioaugmentation of activated sludge.
Solubility
Soluble in water. [6.8 g/L (20°C)].
Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases.
3-Chloroaniline was the only aniline compound to increase plasmaALT/GPT activity at 48 h. It is employed in the bioaugmentation of activated sludge.
Solubility
Soluble in water. [6.8 g/L (20°C)].
Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases.
Specifications
Melting Point | -10°C |
Density | 1.21 |
Boiling Point | 230°C |
Quantity | 500 g |
Odor | Characteristic |
Refractive Index | 1.594 |
UN Number | UN2019 |
Beilstein | 605969 |
Merck Index | 14,2118 |
Solubility Information | Soluble in water. [6.8g/L (20°C)]. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'