missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroaniline 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30320157
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320157 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320157 Supplier Dr. Ehrenstorfer Supplier No. DREXA11351000ME
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Chloroaniline
CAS 108-42-9
Analyte or Component Names 3-Chloroaniline
Shipping Condition Room Temperature
Molecular Formula C6 H6 Cl N
Concentration 100 μg/mL
Certifications/Compliance ISO/IEC 17025
Synonym Aniline, m-chloro- (8CI), 3-Chlorobenzenamine, 1-Amino-3-chlorobenzene, 3-Amino-1-chlorobenzene, 3-Chloro-1-aminobenzene, 3-Chloroaniline, 3-Chlorophenylamine, 5-Chloroaniline, Fast Orange GC Base, NSC 17581, Orange GC base, m-Aminochlorobenzene, m-Chloroaniline, m-Chlorophenylamine
InChI Formula InChI=1S/C6H6ClN/c7-5-2-1-3-6(8)4-5/h1-4H,8H2
Solution Type Single Solution
SMILES Nc1cccc(Cl)c1
IUPAC Name 3-chloroaniline
Solvent or Matrix Methanol
Industry Type Environmental Testing
Molecular Weight (g/mol) 127.57
Quantity 1 mL
Formula Weight 127.0189 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.