missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloroacetophenone, 98%

Product Code. 10765621 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10765621

Brand: Thermo Scientific Acros 134840250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-02-5 | C8H7ClO | 154.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-02-5
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000593
InChI Key UUWJBXKHMMQDED-UHFFFAOYSA-N
Synonym 3'-chloroacetophenone, 1-3-chlorophenyl ethanone, m-chloroacetophenone, ethanone, 1-3-chlorophenyl, 3-chloroacetophenone, 1-3-chlorophenyl ethan-1-one, 3-chloro-acetophenone, acetophenone, 3'-chloro, 1-3-chloro-phenyl-ethanone, 3/'-chloroacetophenone
PubChem CID 14933
IUPAC Name 1-(3-chlorophenyl)ethanone
SMILES CC(=O)C1=CC=CC(Cl)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1900g/mL
Boiling Point 227°C to 229°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5480 to 1.55
Quantity 25 g
Beilstein 07, 282
Merck Index 10, 2082
Specific Gravity 1.19
Formula Weight 154.6
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3'-Chloroacetophenone, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.