missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-N,N-dimethylaniline, 95%

Product Code. 11352586
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352586 5 g 5g
11362586 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352586 Supplier Thermo Scientific Alfa Aesar Supplier No. L08563.06
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 6848-13-1 | C8H10ClN | 155.625 g/mol

It can be used to produce 3-(3-dimethylamino-phenyl)-acrylic acid butyl ester at the temperature of 140°C in the presence of reagent Na2CO3 and solvent N,N-dimethyl-acetamide with the reaction time of 20 hours.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce 3-(3-dimethylamino-phenyl)-acrylic acid butyl ester at the temperature of 140°C in the presence of reagent Na2CO3 and solvent N,N-dimethyl-acetamide with the reaction time of 20 hours.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6848-13-1
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00018456
InChI Key CHHCCYVOJBBCIY-UHFFFAOYSA-N
Synonym n,n-dimethyl-3-chloroaniline, 3-chloro-nn-dimethylaniline, benzenamine, 3-chloro-n,n-dimethyl, m-chloro-n,n-dimethylaniline, benzenamine,3-chloro-n,n-dimethyl, 3-chlorophenyl dimethylamine, pubchem3637, 3-chloranyl-n,n-dimethyl-aniline, 3-n,n-dimethylamino phenyl chloride
PubChem CID 23285
IUPAC Name 3-chloro-N,N-dimethylaniline
SMILES CN(C)C1=CC(=CC=C1)Cl

Specifications

Boiling Point 230°C to 232°C
Refractive Index 1.576
Quantity 5 g
UN Number UN2810
Beilstein 2206152
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 155.63
Percent Purity 95%
Chemical Name or Material 3-Chloro-N,N-dimethylaniline
compliance-icons
Product Identifier
  • 3-Chloro-N,N-dimethylaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3-Chloro-N,N-dimethylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.