missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-6-methoxypyridazine, Thermo Scientific™

Product Code. 11996601 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11996601 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11996601 Supplier Thermo Scientific Acros Supplier No. 148740050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1722-10-7
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.56
MDL Number MFCD00006467
InChI Key XBJLKXOOHLLTPG-UHFFFAOYSA-N
PubChem CID 74403
IUPAC Name 3-chloro-6-methoxypyridazine
SMILES COC1=CC=C(Cl)N=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Synonym pyridazine, 3-chloro-6-methoxy, 3-chloro-6-methoxy-pyridazine, 6-chloro-3-methoxypyridazine, pyridazine,3-chloro-6-methoxy, 6-chloropyridazin-3-yl methyl ether, 3-chloro-6-methoxypyridazine, zlchem 731, acmc-1bvtl, 3-methoxy-6-chloropyridazine, 3-chloro-6-methoxy pyridazine
Formula Weight 144.56
Chemical Name or Material 3-Chloro-6-methoxypyridazine, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.