missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-6-methoxypyridazine, 97%

Product Code. 11462227
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462227 100 g 100g
11442227 5 g 5g
11452227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462227 Supplier Thermo Scientific Alfa Aesar Supplier No. A14173.22
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1722-10-7 | C5H5ClN2O | 144.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1722-10-7
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.56
MDL Number MFCD00006467
InChI Key XBJLKXOOHLLTPG-UHFFFAOYSA-N
PubChem CID 74403
IUPAC Name 3-chloro-6-methoxypyridazine
SMILES COC1=CC=C(Cl)N=N1

Specifications

Melting Point 90°C to 92°C
Quantity 100 g
Beilstein 118854
Synonym pyridazine, 3-chloro-6-methoxy, 3-chloro-6-methoxy-pyridazine, 6-chloro-3-methoxypyridazine, pyridazine,3-chloro-6-methoxy, 6-chloropyridazin-3-yl methyl ether, 3-chloro-6-methoxypyridazine, zlchem 731, acmc-1bvtl, 3-methoxy-6-chloropyridazine, 3-chloro-6-methoxy pyridazine
Formula Weight 144.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-6-methoxypyridazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.