missing translation for 'onlineSavingsMsg'
Learn More

[3-Chloro-5-(trifluoromethyl)-2-pyridyl]methylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10432664
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10432664 1 g 1g
10215624 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10432664 Supplier Thermo Scientific Maybridge Supplier No. SPB06517DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-74-4
Molecular Formula C7H6ClF3N2
Molecular Weight (g/mol) 210.58
MDL Number MFCD00178759
InChI Key JVQYWHGODHSTAM-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl pyridin-2-yl methanamine hydrochloride, 2-aminomethyl-3-chloro-5-trifluoromethyl pyridine hydrochloride, 2-aminomethyl-3-chloro-5-trifluoromethyl-pyridine hydrochloride, 3-chloro-5-trifluoromethyl-2-pyridyl methylamine hydrochloride, 3-chloro-5-trifluoromethyl pyridin-2-yl methanamine hcl, 1-3-chloro-5-trifluoromethyl pyridin-2-yl methanamine hydrochloride, 3-chloro-5-trifluoromethyl pyridin-2-yl methanam, chlorotrifluoromethylpyridinylmethanaminehydrochloride
PubChem CID 11085087
IUPAC Name 1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]methanamine
SMILES NCC1=C(Cl)C=C(C=N1)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 247.05
Percent Purity 97%
Chemical Name or Material [3-Chloro-5-(trifluoromethyl)-2-pyridyl]methylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.