missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorobenzyl alcohol, 98+%

Product Code. 15414807
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414807 5 g 5g
15404807 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414807 Supplier Thermo Scientific Alfa Aesar Supplier No. H32122.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79944-64-2 | C7H6ClFO | 160.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79944-64-2
Molecular Formula C7H6ClFO
Molecular Weight (g/mol) 160.57
MDL Number MFCD03788554
InChI Key VJTJBAMDTCIMOB-UHFFFAOYSA-N
Synonym 3-chloro-5-fluorobenzyl alcohol, 3-chloro-5-fluorophenyl methanol, 3-chloro-5-fluoro-phenyl methanol, rarechem al bd 1247, benzenemethanol,3-chloro-5-fluoro, 3-chloro-5-fluoro phenyl methanol, 3-chloro-5-fluoro-phenyl-methanol, 3-chloro-5-fluorophenyl methan-1-ol
PubChem CID 2734835
IUPAC Name (3-chloro-5-fluorophenyl)methanol
SMILES OCC1=CC(F)=CC(Cl)=C1

Specifications

Refractive Index 1.531
Quantity 5 g
Formula Weight 160.58
Percent Purity ≥98%
Chemical Name or Material 3-Chloro-5-fluorobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.