missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorobenzonitrile, 98%, Thermo Scientific™

Product Code. 10075023 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10075023 25 g Glass bottle 25g
10462713 1 g Glass bottle 1g
10598473 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10075023 Supplier Thermo Scientific Acros Supplier No. 374770250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 327056-73-5
Molecular Formula C7H3ClFN
Molecular Weight (g/mol) 155.56
MDL Number MFCD03407967
InChI Key GIKLRWNRBOLRDV-UHFFFAOYSA-N
Synonym 3-chloro-5-fluoro benzonitrile, 3-chloro-5-fluoro-benzonitrile, benzonitrile, 3-chloro-5-fluoro, 5-chloro-3-fluorobenzenecarbonitrile, 5-chloro-3-fluorobenzonitrile, pubchem4725, acmc-1afkx, 3-fluoro-5-chloro-benzonitrile, 5-fluoro-3-chloro-benzonitrile, buttpark 45\01-07
PubChem CID 2734841
IUPAC Name 3-chloro-5-fluorobenzonitrile
SMILES C1=C(C=C(C=C1F)Cl)C#N

Specifications

Melting Point 53.0°C to 57.0°C
CAS Min % 97.5
Color Yellow
CAS Max % 100.0
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 97.5% min. (GC)
Linear Formula ClC6H3(F)CN
Formula Weight 155.56
Percent Purity 98%
Chemical Name or Material 3-Chloro-5-fluorobenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-5-fluorobenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.