missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorobenzoic acid, 97%, Thermo Scientific™

Product Code. 11416010
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416010 1 g 1g
11426010 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416010 Supplier Thermo Scientific Alfa Aesar Supplier No. H26567.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-5-fluorobenzoic acid is used as a pharmaceutical intermediate. It acts as a protective reagent and is used in chemical research.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25026-64-6
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.555
MDL Number MFCD00973904
InChI Key QFMRANWPGGSNHS-UHFFFAOYSA-N
Synonym 3-chloro-5-fluoro-benzoic acid, 3-fluoro-5-chlorobenzoic acid, 5-chloro-3-fluorobenzoic acid, benzoic acid, 3-chloro-5-fluoro, 3-chloro-5-fluorobenzoicacid, pubchem1373, acmc-209ggm, intermediates-zcf02673, ksc497i9n, rarechem al be 1247
PubChem CID 2734840
IUPAC Name 3-chloro-5-fluorobenzoic acid
SMILES C1=C(C=C(C=C1F)Cl)C(=O)O

Specifications

Quantity 1 g
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 174.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-fluorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.