missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-5-fluorobenzaldehyde, 97%, Thermo Scientific™

Product Code. 10229533 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
20 g
Packaging:
Glass bottle
Unit Size:
20g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10229533 20 g Glass bottle 20g
10451553 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10229533 Supplier Thermo Scientific Acros Supplier No. 374750200
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 90390-49-1
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD02261734
InChI Key QWBYWPJNPKGEBO-UHFFFAOYSA-N
Synonym 3-chloro-5-fluoro-benzaldehyde, 5-chloro-3-fluorobenzaldehyde, benzaldehyde, 3-chloro-5-fluoro, pubchem2890, salor-int l300993-1ea, ksc494e8p, 3-chloro-5-fluorobenzaldehyde
PubChem CID 2734839
IUPAC Name 3-chloro-5-fluorobenzaldehyde
SMILES FC1=CC(Cl)=CC(C=O)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(Cl)CHO
Quantity 20 g
Formula Weight 158.56
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 3-chloro-5-fluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.