missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-pyridinecarboxaldehyde, 97%

Product Code. 10294724 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10294724 5 g Glass bottle 5g
10011754 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10294724 Supplier Thermo Scientific Acros Supplier No. 426550050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72990-37-5 | C6H4ClNO | 141.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72990-37-5
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.56
MDL Number MFCD06200712
InChI Key ZVGDKOQPJCOCLI-UHFFFAOYSA-N
Synonym 3-chloroisonicotinaldehyde, 3-chloropyridine-4-carboxaldehyde, 3-chloro-4-pyridinecarboxaldehyde, 3-chloro-4-pyridinecarbaldehyde, 3-chloro-pyridine-4-carbaldehyde, 3-chloro-4-formylpyridine, 4-pyridinecarboxaldehyde, 3-chloro, acmc-1bgrb, ksc377a6f
PubChem CID 2762995
IUPAC Name 3-chloropyridine-4-carbaldehyde
SMILES C1=CN=CC(=C1C=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 67°C to 68°C
Color White to Yellow
Boiling Point 225°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 141.56
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-pyridinecarboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-4-pyridinecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.