missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylphenylmagnesium bromide, 0.50 M in 2-MeTHF

Product Code. 15490726
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490726 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15490726 Supplier Thermo Scientific Alfa Aesar Supplier No. H54075.AE
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 515158-85-7 | C7H6BrClMg | 229.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.
TRUSTED_SUSTAINABILITY

Specifications

CAS 515158-85-7
Quantity 100 mL
Molecular Formula C7H6BrClMg
MDL Number MFCD11553191
UN Number UN2924
Synonym 3-chloro-4-methylphenylmagnesium bromide, magnesium, bromo 3-chloro-4-methylphenyl, 3-chloro-4-methylphenylmagnesium bromide, 0.50 m in 2-methf, grignard reagent
Solubility Information Reacts with water.
InChI Key MYJXIKNUSSYMAS-UHFFFAOYSA-M
SMILES CC1=CC=C([Mg]Br)C=C1Cl
IUPAC Name magnesium;1-chloro-2-methylbenzene-5-ide;bromide
Molecular Weight (g/mol) 229.78
PubChem CID 24807894
Formula Weight 229.79
Concentration or Composition (by Analyte or Components) 0.50M in 2-MeTHF
Chemical Name or Material 3-Chloro-4-methylphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.