missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-4-methylphenylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 10017759 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10017759 10 g Glass bottle 10g
10421538 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10017759 Supplier Thermo Scientific Acros Supplier No. 430620100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175883-63-3
Molecular Formula C7H8BClO2
Molecular Weight (g/mol) 170.40
MDL Number MFCD04039010
InChI Key YTJUYWRCAZWVSX-UHFFFAOYSA-N
Synonym 3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
PubChem CID 3854610
IUPAC Name (3-chloro-4-methylphenyl)boronic acid
SMILES CC1=CC=C(C=C1Cl)B(O)O

Specifications

Melting Point 228.3°C to 231.5°C
CAS Min % 96.0
Color White
CAS Max % 100.0
Quantity 10 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder of Flakes
Assay Percent Range 96% min. (HPLC)
Formula Weight 170.4
Percent Purity 97%
Chemical Name or Material 3-chloro-4-methylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-4-methylphenylboronic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.