Learn More
3-chloro-4-methylbenzylamine, 97%, Maybridge
Brand: Maybridge TL00917EA
Code : 31
Additional Details : CAS Number : 67952-93-6 Weight : 0.06000kg Transport : UN number : 2735 Chem class : 8 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 10g |
Chemical Identifiers
67952-93-6 | |
155.625 | |
MXIGALIASISPNU-UHFFFAOYSA-N | |
106225 | |
CC1=C(C=C(C=C1)CN)Cl |
C8H10ClN | |
MFCD00014816 | |
3-chloro-4-methylbenzylamine, 3-chloro-4-methylphenyl methanamine, 1-3-chloro-4-methylphenyl methanamine, benzenemethanamine, 3-chloro-4-methyl, 3-chloro-4-methyl benzylamine, 3-chloro-4-methylphenyl methylamine, 3-chloro-4-methyl-benzylamine, rarechem al bw 1348, 4-aminomethyl-2-chlorotoluene, 3-chloro-4-methylbenzenemethanamine | |
(3-chloro-4-methylphenyl)methanamine |
Specifications
10g | |
Amber Glass Bottle | |
97.0 | |
C8H10ClN | |
3-chloro-4-methylbenzylamine, 3-chloro-4-methylphenyl methanamine, 1-3-chloro-4-methylphenyl methanamine, benzenemethanamine, 3-chloro-4-methyl, 3-chloro-4-methyl benzylamine, 3-chloro-4-methylphenyl methylamine, 3-chloro-4-methyl-benzylamine, rarechem al bw 1348, 4-aminomethyl-2-chlorotoluene, 3-chloro-4-methylbenzenemethanamine | |
CC1=C(C=C(C=C1)CN)Cl | |
155.625 | |
155.63 |
3-chloro-4-methylbenzylamine | |
67952-93-6 | |
97% | |
MFCD00014816 | |
MXIGALIASISPNU-UHFFFAOYSA-N | |
(3-chloro-4-methylphenyl)methanamine | |
106225 | |
97% |