missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylbenzoic acid, TRC

Product Code. 30361395
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
250 mg
500 mg
missing translation for 'unitSize'
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361395 500 mg 500mg
30357264 100 mg 100mg
30356710 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30361395 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC587595500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5162-82-3
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
IUPAC Name 3-chloro-4-methylbenzoic acid
SMILES Cc1ccc(cc1Cl)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H7ClO2/c1-5-2-3-6(8(10)11)4-7(5)9/h2-4H,1H3,(H,10,11)
Formula Weight 170.0135
Chemical Name or Material 3-Chloro-4-methylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.