missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-methylaniline, TRC
Click to view available options
Quantity:
500 g
Unit Size:
500g
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-4-methylaniline |
| CAS | 95-74-9 |
| Molecular Formula | C7 H8 Cl N |
| Quantity | 500 g |
| Synonym | p-Toluidine, 3-chloro- (8CI), 3-Chloro-4-methylbenzenamine, 1-Amino-3-chloro-4-methylbenzene, 2-Chloro-4-aminotoluene, 3-Chloro-4-methylaniline, 3-Chloro-4-methylphenylamine, 3-Chloro-p-toluidine, 4-Amino-2-chlorotoluene, 4-Methyl-3-chloroaniline, DRC 1347, NSC 96620 |
| InChI Formula | InChI=1S/C7H8ClN/c1-5-2-3-6(9)4-7(5)8/h2-4H,9H2,1H3 |
| SMILES | Cc1ccc(N)cc1Cl |
| IUPAC Name | 3-chloro-4-methylaniline |
| Molecular Weight (g/mol) | 141.6 |
| Formula Weight | 141.0345 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction