missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methoxybenzaldehyde, 96%, Thermo Scientific™

Product Code. 11395479
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395479 250 mg 250mg
11305489 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395479 Supplier Thermo Scientific Alfa Aesar Supplier No. H50447.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4903-09-7
MDL Number MFCD00016601

Specifications

Molecular Formula C7H8O3
Quantity 250 mg
Sensitivity Air sensitive
Synonym ethyl maltol, 2-ethyl-3-hydroxy-4h-pyran-4-one, 2-ethyl-3-hydroxy-4-pyrone, veltol plus, 3-hydroxy-2-ethyl-4-pyrone, ethylmaltol, 2-ethylpyromeconic acid, 4h-pyran-4-one, 2-ethyl-3-hydroxy, 3-hydroxy-2-ethyl-1,4-pyrone, 2-ethyl pyromeconic acid
InChI Key YIKYNHJUKRTCJL-UHFFFAOYSA-N
SMILES CCC1=C(C(=O)C=CO1)O
IUPAC Name 2-ethyl-3-hydroxypyran-4-one
Molecular Weight (g/mol) 140.14
PubChem CID 21059
Formula Weight 170.6
Percent Purity 96%
Chemical Name or Material 3-Chloro-4-methoxybenzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.