missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog Number 15452139
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
89284-20-8
C5H4ClNO
129.54
MFCD04114248
NKEYLLFRPHHBOQ-UHFFFAOYSA-N
3-chloro-4-hydroxypyridine, 3-chloropyridin-4-ol, 4-pyridinol, 3-chloro, 3-chloropyridin-4 1h-one, 3-chloro-4-hydroxy pyridine, pubchem6680, 3-chloro-4-pyridinol, 3-chloropyridine-4-ol, acmc-20a0ep, 4 1h-pyridinone,3-chloro
2762929
3-chloro-1H-pyridin-4-one
ClC1=CNC=CC1=O
This item is not returnable. View return policy
89284-20-8
C5H4ClNO
129.54
MFCD04114248
NKEYLLFRPHHBOQ-UHFFFAOYSA-N
3-chloro-4-hydroxypyridine, 3-chloropyridin-4-ol, 4-pyridinol, 3-chloro, 3-chloropyridin-4 1h-one, 3-chloro-4-hydroxy pyridine, pubchem6680, 3-chloro-4-pyridinol, 3-chloropyridine-4-ol, acmc-20a0ep, 4 1h-pyridinone,3-chloro
2762929
3-chloro-1H-pyridin-4-one
ClC1=CNC=CC1=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 204°C to 205°C
Quantity 250 mg
Beilstein 1363515
Formula Weight 129.54
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-hydroxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'