missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxypyridine, 98%

Product Code. 15432139
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15432139 1 g 1g
15452139 250 mg 250mg
15442139 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15432139

Brand: Thermo Scientific Alfa Aesar H64783.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 89284-20-8 | C5H4ClNO | 129.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89284-20-8
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.54
MDL Number MFCD04114248
InChI Key NKEYLLFRPHHBOQ-UHFFFAOYSA-N
Synonym 3-chloro-4-hydroxypyridine, 3-chloropyridin-4-ol, 4-pyridinol, 3-chloro, 3-chloropyridin-4 1h-one, 3-chloro-4-hydroxy pyridine, pubchem6680, 3-chloro-4-pyridinol, 3-chloropyridine-4-ol, acmc-20a0ep, 4 1h-pyridinone,3-chloro
PubChem CID 2762929
IUPAC Name 3-chloro-1H-pyridin-4-one
SMILES ClC1=CNC=CC1=O

Specifications

Melting Point 204°C to 205°C
Quantity 1 g
Beilstein 1363515
Formula Weight 129.54
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-hydroxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.