missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4'-fluoropropiophenone, 97%

Product Code. 11494957
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494957 25 g 25g
11484957 5 g 5g
11359499 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494957 Supplier Thermo Scientific Alfa Aesar Supplier No. B21891.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 347-93-3 | C9H8ClFO | 186.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 347-93-3
Molecular Formula C9H8ClFO
Molecular Weight (g/mol) 186.61
MDL Number MFCD00000991
InChI Key AAHQPLJUSLMHHR-UHFFFAOYSA-N
Synonym 3-chloro-4'-fluoropropiophenone, 3-chloro-1-4-fluorophenyl propan-1-one, 3-chloro-1-4-fluorophenyl-1-propanone, 1-propanone, 3-chloro-1-4-fluorophenyl, p-fluoro-, a-chloropropiophenone, pubchem13250, acmc-1ctxg, ksc573a4j, 3-chloro-4'fluoropropiophenone, 3-chloro4'-fluoropropiophenone
PubChem CID 67672
IUPAC Name 3-chloro-1-(4-fluorophenyl)propan-1-one
SMILES C1=CC(=CC=C1C(=O)CCCl)F

Specifications

Melting Point 46°C to 49°C
Boiling Point 100°C to 103°C (3 mmHg)
Quantity 25 g
Beilstein 1866785
Formula Weight 186.61
Percent Purity 97%
Chemical Name or Material 3-Chloro-4'-fluoropropiophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.