missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorophenylhydrazine hydrochloride, 98%

Product Code. 11409472
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409472 100 g 100g
11489462 5 g 5g
11499462 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409472 Supplier Thermo Scientific Alfa Aesar Supplier No. A10175.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 175135-74-7 | C6H7Cl2FN2 | 197.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175135-74-7
Molecular Formula C6H7Cl2FN2
Molecular Weight (g/mol) 197.034
MDL Number MFCD00052267
InChI Key DWCDDXINAZHRLK-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorophenylhydrazine hydrochloride, 3-chloro-4-fluorophenyl hydrazine hydrochloride, 1-3-chloro-4-fluorophenyl hydrazine hydrochloride, hydrazine, 3-chloro-4-fluorophenyl-, monohydrochloride, pubchem3507, acmc-1c5q6, labotest-bb lt01147590, ksc494o5j, 3-chloro-4-fluorophenylhydrazinehcl, 3-chloro-4-fluorophenylhydrazine hcl
PubChem CID 44181777
IUPAC Name (3-chloro-4-fluorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=C(C=C1NN)Cl)F.Cl

Specifications

Melting Point ∼210°C (decomposition)
Quantity 100 g
Formula Weight 197.04
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluorophenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.