missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorophenylboronic acid, 97%, ACROS Organics™

Product Code. 10560542 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10560542

Brand: Thermo Scientific Acros 367680050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 144432-85-9
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.362
MDL Number MFCD00051800
InChI Key WJDZZXIDQYKVDG-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorobenzeneboronic acid, 3-chloro-4-fluorophenyl boronic acid, 3-chloro-4-fluorophenyl boranediol, 4-fluoro-3-chlorophenylboronic acid, 3-chloro-4-fluoro phenyl boronic acid, 3-chloro-4-fluoro-phenyl boronic acid, boronic acid, 3-chloro-4-fluorophenyl, b-3-chloro-4-fluorophenyl-boronic acid, pubchem1780
PubChem CID 2734660
IUPAC Name (3-chloro-4-fluorophenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)F)Cl)(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 174.37
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 3-Chloro-4-fluorophenylboronic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.