missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-4-fluorophenyl isocyanate, 98%, Thermo Scientific™

Product Code. 10190222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10190222 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10190222 Supplier Thermo Scientific Acros Supplier No. 366790010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

CAS 50529-33-4
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3770g/mL
Boiling Point 44°C (17.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C7H3ClFNO
Linear Formula Cl(F)C6H3NCO
Refractive Index 1.5395 to 1.5415
MDL Number MFCD00037037
Quantity 1 g
Specific Gravity 1.377
Synonym 3-chloro-4-fluorophenyl isocyanate, 3-chloro-4-fluorophenylisocyanate, benzene, 2-chloro-1-fluoro-4-isocyanato, 3-chloro-4-fluorobenzenisocyanate, pubchem5024, acmc-20an6r, 3-chloro-4-fluoro-phenylisocyanate, 3-chloro-4-fluorophenyl-isocyanate, 3-chloro-4-fluoro phenyl isocyanate
InChI Key XVIPJBUXMFLHSI-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1N=C=O)Cl)F
IUPAC Name 2-chloro-1-fluoro-4-isocyanatobenzene
Molecular Weight (g/mol) 171.56
PubChem CID 2734632
Formula Weight 171.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Chloro-4-fluorophenyl isocyanate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-4-fluorophenyl isocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.