Learn More
Thermo Scientific Maybridge 3-Chloro-4-fluorobenzylamine, 1g, 97%
Brand: Thermo Scientific Maybridge TL00177DA
3-Chloro-4-fluorobenzylamine, 1g, 97%
Code : 31
Additional Details : CAS Number : 72235-56-4 Weight : 0.05100kg Transport : UN number : 1760 Chem class : 8 Pack group : III