missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzyl bromide, 97%, Thermo Scientific™

Catalog Number 11364036
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
192702-01-5
C7H5BrClF
223.47
MFCD01631551
GGTQWWTYUKXFPP-UHFFFAOYSA-N
3-chloro-4-fluorobenzyl bromide, 4-bromomethyl-2-chloro-1-fluorobenzene, alpha-bromo-3-chloro-4-fluorotoluene, 1-bromomethyl-3-chloro-4-fluorobenzene, 3-chloro-4-fluorobenzylbromide, 3-chloro-4-fluoro benzyl bromide, benzene, 4-bromomethyl-2-chloro-1-fluoro, pubchem2130, acmc-209ewf, 4-bromomethyl-2-chloro-1-fluoro-benzene
2757542
4-(bromomethyl)-2-chloro-1-fluorobenzene
FC1=CC=C(CBr)C=C1Cl
This item is not returnable. View return policy
192702-01-5
C7H5BrClF
223.47
MFCD01631551
GGTQWWTYUKXFPP-UHFFFAOYSA-N
3-chloro-4-fluorobenzyl bromide, 4-bromomethyl-2-chloro-1-fluorobenzene, alpha-bromo-3-chloro-4-fluorotoluene, 1-bromomethyl-3-chloro-4-fluorobenzene, 3-chloro-4-fluorobenzylbromide, 3-chloro-4-fluoro benzyl bromide, benzene, 4-bromomethyl-2-chloro-1-fluoro, pubchem2130, acmc-209ewf, 4-bromomethyl-2-chloro-1-fluoro-benzene
2757542
4-(bromomethyl)-2-chloro-1-fluorobenzene
FC1=CC=C(CBr)C=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 63°C to 65°C (0.6 mmHg)
Refractive Index 1.568
Quantity 250 mg
UN Number UN3265
Beilstein 2435145
Formula Weight 223.48
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-fluorobenzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'