missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzotrifluoride, 98%

Product Code. 11412658
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412658 5 g 5g
11422658 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412658 Supplier Thermo Scientific Alfa Aesar Supplier No. A16670.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 78068-85-6 | C7H3ClF4 | 198.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78068-85-6
Molecular Formula C7H3ClF4
Molecular Weight (g/mol) 198.54
MDL Number MFCD00042207
InChI Key BKHVEYHSOXVAOP-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorobenzotrifluoride, 2-chloro-1-fluoro-4-trifluoromethyl benzene, benzene, 2-chloro-1-fluoro-4-trifluoromethyl, 3-cholor-4-fluorobenzo-trifluoride, 3-chloro-4-fluoro benzotrifluoride, 1-chloro-2-fluoro-5-trifluoromethyl benzene, 3-chloro-4-fluoro-1-trifluoromethyl benzene, benzene, chloro-1-fluoro-4-trifluoromethyl, 3-chloro-alpha,alpha,alpha,4-tetrafluorotoluene, 3-chloro-4-fluoro-alpha,alpha,alpha-trifluorotoluene
PubChem CID 157153
IUPAC Name 2-chloro-1-fluoro-4-(trifluoromethyl)benzene
SMILES FC1=CC=C(C=C1Cl)C(F)(F)F

Specifications

Density 1.49
Boiling Point 137°C
Flash Point 42°C (108°F)
Refractive Index 1.4355
Quantity 5 g
UN Number UN1993
Beilstein 4309386
Formula Weight 198.55
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluorobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.