missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15477266
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
635305-46-3
C12H15BClFO2
256.508
PFRXRUVRRFJIBF-UHFFFAOYSA-N
2-3-chloro-4-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-chloro-4-fluorobenzeneboronic acid pinacol ester, 2-3-chloro-4-fluorophenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 1,3,2-dioxaborolane, 2-3-chloro-4-fluorophenyl-4,4,5,5-tetramethyl
10015303
2-(3-chloro-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)Cl
This item is not returnable. View return policy
635305-46-3
C12H15BClFO2
256.508
PFRXRUVRRFJIBF-UHFFFAOYSA-N
2-3-chloro-4-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-chloro-4-fluorobenzeneboronic acid pinacol ester, 2-3-chloro-4-fluorophenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 1,3,2-dioxaborolane, 2-3-chloro-4-fluorophenyl-4,4,5,5-tetramethyl
10015303
2-(3-chloro-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 256.51
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Chloro-4-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'