missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluoroaniline, 98%

Product Code. 11423824
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423824 25 g 25g
11433824 100 g 100g
11443824 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423824 Supplier Thermo Scientific Alfa Aesar Supplier No. A11858.14
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 367-21-5 | C6H5ClFN | 145.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 367-21-5
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00007767
InChI Key YSEMCVGMNUUNRK-UHFFFAOYSA-N
Synonym benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin
PubChem CID 9708
IUPAC Name 3-chloro-4-fluoroaniline
SMILES NC1=CC=C(F)C(Cl)=C1

Spécification

Melting Point 44°C to 47°C
Density 1.42
Quantity 25 g
Odor Characteristic
UN Number UN2811
Beilstein 1562786
Boiling Point 227°C to 230°C
Formula Weight 145.56
Percent Purity ≥98%
Chemical Name or Material 3-Chloro-4-fluoroaniline
compliance-icons
Identifiant produit
  • 3-Chloro-4-fluoroaniline
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Informations supplémentaires
  • MIXTURE LIST-Contain: 3-Chloro-4-fluoroaniline

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.