missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluoroaniline, 98%

Product Code. 10686972 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10686972 25 g Glass bottle 25g
10722831 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10686972 Supplier Thermo Scientific Acros Supplier No. 211530250
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 367-21-5 | C6H5ClFN | 145.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-21-5
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00007767
InChI Key YSEMCVGMNUUNRK-UHFFFAOYSA-N
Synonym benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin
PubChem CID 9708
IUPAC Name 3-chloro-4-fluoroaniline
SMILES NC1=CC=C(F)C(Cl)=C1

Specifications

Melting Point 42°C to 47°C
CAS Min % 97.5
Color Beige to Brown
CAS Max % 100.0
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Assay Percent Range 98%
Linear Formula ClC6H3(F)NH2
Beilstein 12, I, 309
Boiling Point 230°C
Solubility Information Solubility in water: 10g/L water 520°C). Other solubilities: soluble in organic solvents
Formula Weight 145.56
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluoroaniline, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-4-fluoroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.