missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluoroaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7081748
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
367-21-5
C6H5ClFN
145.56
MFCD00007767
YSEMCVGMNUUNRK-UHFFFAOYSA-N
benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin
9708
3-chloro-4-fluoroaniline
NC1=CC=C(F)C(Cl)=C1
This item is not returnable. View return policy
367-21-5
C6H5ClFN
145.56
MFCD00007767
YSEMCVGMNUUNRK-UHFFFAOYSA-N
benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin
9708
3-chloro-4-fluoroaniline
NC1=CC=C(F)C(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 44°C to 47°C
Density 1.42
Boiling Point 227°C to 230°C
Quantity 25 g
Odor Characteristic
UN Number UN2811
Beilstein 1562786
Formula Weight 145.56
Percent Purity ≥98%
Chemical Name or Material 3-Chloro-4-fluoroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'