missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(ethylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15440436
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
850589-40-1
C9H11BClNO3
227.45
MFCD07363767
HJDYETAQUCUPFP-UHFFFAOYSA-N
3-chloro-4-ethylcarbamoyl phenyl boronic acid, 3-chloro-4-n-ethylcarbamoyl phenylboronic acid, 3-chloro-4-n-ethylcarbamoyl benzeneboronic acid, 3-chloro-4-ethylcarbamoyl phenylboronic acid, boronic acid,b-3-chloro-4-ethylamino carbonyl phenyl, 3-chloro-4-n-ethyl carbamoyl phenylboronic acid, 3-chloranyl-4-ethylcarbamoyl phenyl boronic acid
44119416
[3-chloro-4-(ethylcarbamoyl)phenyl]boronic acid
CCNC(=O)C1=C(Cl)C=C(C=C1)B(O)O
This item is not returnable. View return policy
850589-40-1
C9H11BClNO3
227.45
MFCD07363767
HJDYETAQUCUPFP-UHFFFAOYSA-N
3-chloro-4-ethylcarbamoyl phenyl boronic acid, 3-chloro-4-n-ethylcarbamoyl phenylboronic acid, 3-chloro-4-n-ethylcarbamoyl benzeneboronic acid, 3-chloro-4-ethylcarbamoyl phenylboronic acid, boronic acid,b-3-chloro-4-ethylamino carbonyl phenyl, 3-chloro-4-n-ethyl carbamoyl phenylboronic acid, 3-chloranyl-4-ethylcarbamoyl phenyl boronic acid
44119416
[3-chloro-4-(ethylcarbamoyl)phenyl]boronic acid
CCNC(=O)C1=C(Cl)C=C(C=C1)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 142°C to 146°C
Quantity 1 g
Formula Weight 227.45
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(ethylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'