missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-ethoxyphenylboronic acid, 98%, Thermo Scientific™

Catalog Number 15254368
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
279261-81-3
C8H10BClO3
200.43
QZMQZKMQXURQNC-UHFFFAOYSA-N
3-chloro-4-ethoxyphenyl boronic acid, 3-chloro-4-ethoxybenzeneboronic acid, 3-chloro-4-ethoxypheylboronic acid, 3-chloro-4-ethoxy-phenyl boronic acid, acmc-209gzs, 3-chloro-4-ethoxyphenyl boronicacid, 3-chloranyl-4-ethoxy-phenyl boronic acid, boronic acid,b-3-chloro-4-ethoxyphenyl, 3-chloro-4-ethoxyphenylboronicacid containsvaryingamountsofanhydride
3717658
(3-chloro-4-ethoxyphenyl)boronic acid
B(C1=CC(=C(C=C1)OCC)Cl)(O)O
This item is not returnable. View return policy
279261-81-3
C8H10BClO3
200.43
QZMQZKMQXURQNC-UHFFFAOYSA-N
3-chloro-4-ethoxyphenyl boronic acid, 3-chloro-4-ethoxybenzeneboronic acid, 3-chloro-4-ethoxypheylboronic acid, 3-chloro-4-ethoxy-phenyl boronic acid, acmc-209gzs, 3-chloro-4-ethoxyphenyl boronicacid, 3-chloranyl-4-ethoxy-phenyl boronic acid, boronic acid,b-3-chloro-4-ethoxyphenyl, 3-chloro-4-ethoxyphenylboronicacid containsvaryingamountsofanhydride
3717658
(3-chloro-4-ethoxyphenyl)boronic acid
B(C1=CC(=C(C=C1)OCC)Cl)(O)O
Melting Point 69.0°C to 70.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 200.43
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-ethoxyphenylboronic acid
compliance-icons
Product Identifier
  • 3-Chloro-4-ethoxyphenylboronic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'