missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(1-piperidinylcarbonyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15430026
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850589-50-3
Molecular Formula C12H15BClNO3
Molecular Weight (g/mol) 267.516
MDL Number MFCD07363775
InChI Key AQTGPCIHLDJZJF-UHFFFAOYSA-N
Synonym 3-chloro-4-piperidine-1-carbonyl phenylboronic acid, 3-chloro-4-piperidine-1-carbonyl phenyl boronic acid, 3-chloro-4-piperidin-1-ylcarbonyl benzeneboronic acid, boronic acid,b-3-chloro-4-1-piperidinylcarbonyl phenyl, 1-4-borono-2-chlorobenzoyl piperidine, n-piperidinyl 2-chloro-4-boronobenzamide, 4-borono-2-chlorophenyl piperidin-1-yl methanone, 3-chloro-4-1-piperidinylcarbonyl benzeneboronic acid, b-3-chloro-4-1-piperidinylcarbonyl phenyl-boronic acid
PubChem CID 44119648
IUPAC Name [3-chloro-4-(piperidine-1-carbonyl)phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)C(=O)N2CCCCC2)Cl)(O)O

Specifications

Melting Point 160°C to 163°C
Quantity 250 mg
Formula Weight 267.52
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-(1-piperidinylcarbonyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'