missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-3-methyl-1-butyne, TRC

Product Code. 30362604
Change view
Click to view available options
Quantity:
5 g
Product Code. Quantity
30362604 5 g
1 options
This item is not returnable. View return policy

Product Code. 30362604

Brand: Toronto Research Chemicals TRCC3690905G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Chloro-3-methyl-1-butyne
CAS 1111-97-3
Molecular Formula C5H7Cl
Quantity 5 g
Synonym 2-Chloro-2-methyl-3-butyne, 2-Methyl-2-chloro-3-butyne, 2-Methyl-3-butyn-2-yl chloride, 3-Chloro-3-methylbutyne, 3-Methyl-1-butyn-3-yl chloride, 3-Methyl-3-chloro-1-butyne, 3-Methyl-3-chlorobutyne, NSC 16173, alpha,alpha-Dimethylpropargyl Chloride
InChI Formula InChI=1S/C5H7Cl/c1-4-5(2,3)6/h1H,2-3H3
SMILES CC(C)(Cl)C#C
IUPAC Name 3-chloro-3-methylbut-1-yne
Molecular Weight (g/mol) 102.56
Formula Weight 102.0236
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.