missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-chloro-2-methylphenyl isothiocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Synonym | 3-chloro-2-methylphenyl isothiocyanate, isothiocyanic acid, 3-chloro-o-tolyl ester, 3-chloro-2-methylphenylisothiocyanate, benzene,1-chloro-3-isothiocyanato-2-methyl, 3-chloro-2-methylbenzenisothiocyanate, pubchem10477, 3-chloro-2-methylphenyl-isothiocyanate, 1-chloranyl-3-isothiocyanato-2-methyl-benzene, benzene, 1-chloro-3-isothiocyanato-2-methyl |
Formula Weight | 183.66 |
Percent Purity | 97% |
Quantity | 25 g |
Chemical Name or Material | 3-Chloro-2-methylphenyl isothiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'