missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylphenyl isocyanate, 99%

Product Code. 11313006
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11313006

Brand: Thermo Scientific Alfa Aesar L10802.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40397-90-8 | C8H6ClNO | 167.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40397-90-8
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD00037036
InChI Key NGQMCUWZGVMILT-UHFFFAOYSA-N
Synonym 3-chloro-2-methylphenyl isocyanate, 3-chloro-2-methylphenylisocyanate, 1-chloro-3-isocyanato-2-methyl-benzene, 3-chloro-2-methylbenzenisocyanate, acmc-1aqe4, 2-chloro-6-isocyanatotoluene, benzene,1-chloro-3-isocyanato-2-methyl, 1-chloranyl-3-isocyanato-2-methyl-benzene, benzene, 1-chloro-3-isocyanato-2-methyl
PubChem CID 2735770
IUPAC Name 1-chloro-3-isocyanato-2-methylbenzene
SMILES CC1=C(C=CC=C1Cl)N=C=O

Specifications

Density 1.247
Boiling Point 94°C to 95°C (9 mmHg)
Quantity 1 g
Refractive Index 1.559
UN Number UN2206
Beilstein 2691111
Formula Weight 167.6
Percent Purity 99%
Chemical Name or Material 3-Chloro-2-methylphenyl isocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.