missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylanisole, 97%

Product Code. 11455826
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455826 100 g 100g
11435826 5 g 5g
11445826 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455826 Supplier Thermo Scientific Alfa Aesar Supplier No. B22203.22
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3260-88-6 | C8H9ClO | 156.61 g/mol

Arene-manganese tricarbonyl complexes are prepared using 3-Chloro-2-methylanisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Arene-manganese tricarbonyl complexes are prepared using 3-Chloro-2-methylanisole.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3260-88-6
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00070772
InChI Key LTVRGAWOEOKGJZ-UHFFFAOYSA-N
Synonym 3-chloro-2-methylanisole, 2-methyl-3-chloroanisole, 2-chloro-6-methoxytoluene, benzene, 1-chloro-3-methoxy-2-methyl, 2-methoxy-6-chlorotoluene, 1-chloro-3-methoxy-2-methyl-benzene, pubchem2664, acmc-1cq3o, ltvrgawoeokgjz-uhfffaoysa
PubChem CID 76749
IUPAC Name 1-chloro-3-methoxy-2-methylbenzene
SMILES COC1=CC=CC(Cl)=C1C

Specifications

Melting Point -3°C
Density 1.16
Boiling Point 212°C to 215°C
Flash Point 92°C (197°F)
Refractive Index 1.5395
Quantity 100 g
UN Number UN1993
Beilstein 1862220
Formula Weight 156.61
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-methylanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.