missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-hydroxypropyltrimethyl Ammonium Chloride (60% in Water), TRC

Product Code. 30364505
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30364505 10 g
30358775 1 g
30358139 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364505 Supplier Toronto Research Chemicals Supplier No. TRCC36808510G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3327-22-8
Molecular Formula C6H15Cl2NO
Molecular Weight (g/mol) 188.1
Synonym (2-Hydroxy-3-chloropropyl)trimethylammonium chloride, (3-Chloro-2- hydroxypropyl)-N,N,N-trimethylammonium chloride, 1-Chloro-2-hydroxypropane-3-trimethylammonium chloride, 1-Chloro-3-(trimethylammonio)-2- propanol chloride, 1-Chloro-3-trimethylammonium-2-propanol chloride, COPA 60S, CR 2000, CR 2000 (fixative), Catiomaster C, Catiomaster C 60, Cation Master C, Dextrosil, Dextrosil KA, Dowquat 188, N-(3-Chloro-2-hydroxypropyl)-N,N,N-trimethylammonium chloride, NSC 51216, NT 21, QUAB, QUAB 188, QUAB 218, Quat 188, Reagens CFZ, Reagens S, Reagens S-CFZ, Trimethyl(2-hydroxy-3-chloropropyl)ammonium chloride, Verolan KAF, Vircotone ECO
IUPAC Name (3-chloro-2-hydroxypropyl)-trimethylazanium;chloride
SMILES [Cl-].C[N+](C)(C)CC(O)CCl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H15ClNO.ClH/c1-8(2,3)5-6(9)4-7;/h6,9H,4-5H2,1-3H3;1H/q+1;/p-1
Formula Weight 187.0531
Concentration or Composition (by Analyte or Components) 0.6
Chemical Name or Material 3-Chloro-2-hydroxypropyltrimethyl Ammonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.