missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluoroaniline, 97%

Product Code. 15488017
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488017 5 g 5g
15408027 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488017 Supplier Thermo Scientific Alfa Aesar Supplier No. H51144.06
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2106-04-9 | C8H11Cl2N5 | 248.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2106-04-9
MDL Number MFCD00069415

Specifications

Density 1.324
Quantity 5 g
Odor Characteristic
Refractive Index 1.564
Molecular Formula C6H5ClFN
UN Number UN2810
Synonym 2-fluoro-3-chloroaniline, benzenamine, 3-chloro-2-fluoro, 3-chloro-2-fluorophenylamine, pubchem1529, 3,4-chlorofluoroaniline, 3-chloro-2 fluoroaniline, 2-fluoro-3-chloro-aniline, 3-chloro-2-fluoro aniline, 3-chloro-2-fluoro-aniline, ksc493k4l
Boiling Point 214°C
InChI Key XWBTZHDDWRNOQH-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)F)N
IUPAC Name 3-chloro-2-fluoroaniline
Molecular Weight (g/mol) 145.56
PubChem CID 75014
Formula Weight 145.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.