missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluoroaniline, 97%

Product Code. 15408027
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408027 100 g 100g
15488017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408027 Supplier Thermo Scientific Alfa Aesar Supplier No. H51144.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2106-04-9 | C8H11Cl2N5 | 248.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2106-04-9
MDL Number MFCD00069415

Specifications

Density 1.324
Quantity 100 g
Odor Characteristic
Refractive Index 1.564
Molecular Formula C8H11Cl2N5
UN Number UN2810
Synonym 1-3-chlorophenyl biguanide hydrochloride, m-chlorophenylbiguanide hydrochloride, 1-m-chlorophenyl-biguanide hydrochloride, m-cpbg hydrochloride, 1-m-chlorophenyl biguanide hydrochloride, 1-carbamimidamido-n-3-chlorophenyl methanimidamide hydrochloride, imidodicarbonimidic diamide, n-3-chlorophenyl-, monohydrochloride, n-m-chlorophenylbiguanide hydrochloride, biguanide, 1-m-chlorophenyl-, monohydrochloride, n-3-chlorophenyl imidodicarbonimidic diamide hydrochloride
Boiling Point 214°C
InChI Key FOWAIJYHRWFTHR-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)Cl)N=C(N)N=C(N)N.Cl
IUPAC Name 2-(3-chlorophenyl)-1-(diaminomethylidene)guanidine;hydrochloride
Molecular Weight (g/mol) 248.11
PubChem CID 2730228
Formula Weight 145.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.