missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-(chloromethyl)-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. p-3522488
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10357033 1 g Amber Glass Bottle 1g
10613835 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10357033 Supplier Thermo Scientific Maybridge Supplier No. SPB05830DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-52-8
Molecular Formula C7H4Cl2F3N
Molecular Weight (g/mol) 230.011
MDL Number MFCD00173852
InChI Key NUSXEDNPFNWFDQ-UHFFFAOYSA-N
PubChem CID 22058750
IUPAC Name 3-chloro-2-(chloromethyl)-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC(=C1Cl)CCl)C(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-chloro-2-chloromethyl-5-trifluoromethyl pyridine, 3-chloranyl-2-chloromethyl-5-trifluoromethyl pyridine, pyridine,3-chloro-2-chloromethyl-5-trifluoromethyl, pyridine, 3-chloro-2-chloromethyl-5-trifluoromethyl
Formula Weight 230.02
Percent Purity 95%
Chemical Name or Material 3-Chloro-2-(chloromethyl)-5-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.