missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-2-chloromethyl-1-propene, 96%

Product Code. 10185500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10185500 5 g Glass bottle 5g
10532461 25 g Glass bottle 25g
10135743 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10185500 Supplier Thermo Scientific Acros Supplier No. 277070050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1871-57-4 | C4H6Cl2 | 124.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1871-57-4
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 124.99
MDL Number MFCD00000952
InChI Key XJFZOSUFGSANIF-UHFFFAOYSA-N
Synonym 3-chloro-2-chloromethyl prop-1-ene, 3-chloro-2-chloromethyl-1-propene, 1,1-bis chloromethyl ethylene, methallyl dichloride, 1-propene, 3-chloro-2-chloromethyl, 2-chloromethyl-3-chloropropene, 1,3-dichloro-2-methylenepropane, 3-chloro-2-chloromethyl propene, 2-methylene-1,3-dichloropropane, 2-methylenepropane-1,3-dichloride
PubChem CID 15859
IUPAC Name 3-chloro-2-(chloromethyl)prop-1-ene
SMILES ClCC(=C)CCl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 14°C
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 138°C to 140°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH2Cl)2
Refractive Index 1.4750 to 1.479
Quantity 5 g
Beilstein 01, II, 181
Specific Gravity 1.08
Solubility Information Solubility in water: insoluble
Formula Weight 125
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-chloro-2-chloromethyl-1-propene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Chloro-2-chloromethyl-1-propene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.