missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloro-2-bromopropiophenone, TRC

Product Code. 30133534
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133534 250 mg 250mg
30138563 1 g 1g
17960855 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133534 Supplier Toronto Research Chemicals Supplier No. TRCC365035250MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34911-51-8
Molecular Formula C9 H8 Br Cl O
Molecular Weight (g/mol) 247.52
Synonym 2-Bromo-1-(3-chlorophenyl)-1-propanone, Propiophenone, 2-bromo-3'-chloro- (7CI), 2-Bromo-1-(3-chlorophenyl)propan-1-one, 2-Bromo-3'-chloropropiophenone, α-Bromo-3-chloropropiophenone, α-Bromo-m-chloropropiophenone
IUPAC Name 2-bromo-1-(3-chlorophenyl)propan-1-one
SMILES CC(Br)C(=O)c1cccc(Cl)c1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H8BrClO/c1-6(10)9(12)7-3-2-4-8(11)5-7/h2-6H,1H3
Formula Weight 245.9447
Chemical Name or Material 2-Bromo-1-(3-chlorophenyl)propan-1-one (2-Bromo-3'-chloropropiophenone)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.