missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,6-dimethylaniline, 99%

Product Code. 11435876
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11435876

Brand: Thermo Scientific Alfa Aesar B22232.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26829-77-6 | C8H10ClN | 155.625 g/mol

3-cyano- 2,4-dimethylchlorobenzene was obtained from 3-Chloro-2,6-dimethylaniline which upon hydrolysis gives acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-cyano- 2,4-dimethylchlorobenzene was obtained from 3-Chloro-2,6-dimethylaniline which upon hydrolysis gives acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26829-77-6
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD02683086
InChI Key FGMXFOTYCHZCLA-UHFFFAOYSA-N
Synonym 3-chloro-2,6-dimethyl-phenylamine, acmc-20an7h, 3-chloro-2,6-xylidine, 3-chloro-2,6-dimethylphenylamine, 3-chloranyl-2,6-dimethyl-aniline, benzenamine,3-chloro-2,6-dimethyl
PubChem CID 7023016
IUPAC Name 3-chloro-2,6-dimethylaniline
SMILES CC1=C(C(=C(C=C1)Cl)C)N

Specifications

Melting Point 30°C to 32°C
Boiling Point 136°C to 138°C (17 mmHg)
Quantity 50 g
Solubility Information Slightly soluble in water.
Formula Weight 155.63
Percent Purity 99%
Chemical Name or Material 3-Chloro-2,6-dimethylaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.