missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,6-difluorobenzamide, 97+%, Thermo Scientific™

Product Code. 11401057
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261762-41-8
Molecular Formula C7H4ClF2NO
Molecular Weight (g/mol) 191.562
MDL Number MFCD01631386
InChI Key TUTOELHQVJTZTO-UHFFFAOYSA-N
Synonym acmc-1cfko, benzamide,3-chloro-2,6-difluoro
PubChem CID 2773517
IUPAC Name 3-chloro-2,6-difluorobenzamide
SMILES C1=CC(=C(C(=C1F)C(=O)N)F)Cl

Specifications

Melting Point 133°C to 136°C
Quantity 1 g
Formula Weight 191.56
Percent Purity ≥97%
Chemical Name or Material 3-Chloro-2,6-difluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'