Learn More
3-Chloro-2,4-pentanedione, 98%, Thermo Scientific Chemicals
£24.60 - £773.28
Chemical Identifiers
CAS | 1694-29-7 |
---|---|
Molecular Formula | C5H7ClO2 |
Molecular Weight (g/mol) | 134.56 |
MDL Number | MFCD00009651 |
InChI Key | VLRGXXKFHVJQOL-UHFFFAOYSA-N |
PubChem CID | 74328 |
IUPAC Name | 3-chloropentane-2,4-dione |
SMILES | CC(=O)C(C(=O)C)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10773591
|
Thermo Scientific Acros
210670050 |
5 mL |
£24.60
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10256550
|
Thermo Scientific Acros
210671000 |
100 mL |
£49.90
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10636354
|
Thermo Scientific Acros
210675000 |
500 mL | |||||||
10509063
|
Thermo Scientific Acros
210670025 |
2.5 L | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1694-29-7 | |
134.56 | |
VLRGXXKFHVJQOL-UHFFFAOYSA-N | |
3-chloropentane-2,4-dione |
C5H7ClO2 | |
MFCD00009651 | |
74328 | |
CC(=O)C(C(=O)C)Cl |
Specifications
1694-29-7 | |
100.0 | |
Brown to Yellow | |
49°C to 52°C (18.0 mmHg) | |
Authentic | |
Glass bottle | |
CH3COCH(Cl)COCH3 | |
MFCD00009651 | |
1.179 | |
Solubility in water: 7.65g/L (20°C). Other solubilities: soluble in organic solvents | |
CC(=O)C(C(=O)C)Cl | |
134.56 | |
134.56 | |
Liquid |
97.5 | |
-15°C | |
1.1790g/mL | |
56°C | |
97.5% min. (GC) | |
C5H7ClO2 | |
1.4825 to 1.4845 | |
01, 785 | |
3-chloro-2,4-pentanedione, 3-chloroacetylacetone, 2,4-pentanedione, 3-chloro, 1-acetyl-1-chloroacetone, 3-chloro 2,4-pentanedione, 3-chloro.2,4-pentanedione, 3-chloro-pentane-2,4-dione, 2,4-pentanedione,3-chloro, 3-chloro-2,4-pentanedione, produced by wacker chemie ag, burghausen, germany gc | |
VLRGXXKFHVJQOL-UHFFFAOYSA-N | |
3-chloropentane-2,4-dione | |
74328 | |
98% | |
3-Chloro-2, 4-pentanedione, 98% |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 216-902-4
missing translation for 'tsca' : TSCA
- 3-Chloro-2,4-pentanedione
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only