missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,4-pentanedione, 98%

Product Code. 10256550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10256550 100 mL 100mL
10773591 5 mL 5mL
10636354 500 mL 500mL
10509063 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10256550 Supplier Thermo Scientific Acros Supplier No. 210671000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1694-29-7 | C5H7ClO2 | 134.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1694-29-7
Molecular Formula C5H7ClO2
Molecular Weight (g/mol) 134.56
MDL Number MFCD00009651
InChI Key VLRGXXKFHVJQOL-UHFFFAOYSA-N
PubChem CID 74328
IUPAC Name 3-chloropentane-2,4-dione
SMILES CC(=O)C(C(=O)C)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -15°C
Color Brown to Yellow
Density 1.1790g/mL
Boiling Point 49°C to 52°C (18.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH(Cl)COCH3
Refractive Index 1.4825 to 1.4845
Quantity 100 mL
Beilstein 01, 785
Specific Gravity 1.179
Synonym 3-chloro-2,4-pentanedione, 3-chloroacetylacetone, 2,4-pentanedione, 3-chloro, 1-acetyl-1-chloroacetone, 3-chloro 2,4-pentanedione, 3-chloro.2,4-pentanedione, 3-chloro-pentane-2,4-dione, 2,4-pentanedione,3-chloro, 3-chloro-2,4-pentanedione, produced by wacker chemie ag, burghausen, germany gc
Solubility Information Solubility in water: 7.65g/L (20°C). Other solubilities: soluble in organic solvents
Formula Weight 134.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Chloro-2, 4-pentanedione, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Chloro-2,4-pentanedione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.